Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCGCCTGTCTTGTGGGGACCCACC[A/G]ATGTCAGTGGCCAGCTGCTTGGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606978 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COG7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COG7 - component of oligomeric golgi complex 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153603.3 | 2342 | Silent Mutation | ATC,ATT | I,I 715 | NP_705831.1 | |
XM_017023870.1 | 2342 | Silent Mutation | ATC,ATT | I,I 650 | XP_016879359.1 |