Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGGTGGGGATGGCACCGCCCTAC[C/G]GCCGAGAGAGTTGAAGCTGCACCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603895 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM234A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM234A - family with sequence similarity 234 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284497.1 | 2261 | Intron | NP_001271426.1 | |||
NM_032039.3 | 2261 | Intron | NP_114428.1 | |||
XM_005255622.1 | 2261 | Intron | XP_005255679.1 | |||
XM_005255623.2 | 2261 | Intron | XP_005255680.1 | |||
XM_006720957.1 | 2261 | Intron | XP_006721020.1 | |||
XM_011522691.1 | 2261 | Intron | XP_011520993.1 | |||
XM_011522692.1 | 2261 | Intron | XP_011520994.1 | |||
XM_017023760.1 | 2261 | Intron | XP_016879249.1 | |||
XM_017023761.1 | 2261 | Intron | XP_016879250.1 | |||
XM_017023762.1 | 2261 | Intron | XP_016879251.1 | |||
XM_017023763.1 | 2261 | Intron | XP_016879252.1 | |||
XM_017023764.1 | 2261 | Intron | XP_016879253.1 | |||
XM_017023765.1 | 2261 | Intron | XP_016879254.1 |
RGS11 - regulator of G-protein signaling 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286485.1 | 2261 | UTR 3 | NP_001273414.1 | |||
NM_001286486.1 | 2261 | UTR 3 | NP_001273415.1 | |||
NM_003834.2 | 2261 | UTR 3 | NP_003825.1 | |||
NM_183337.2 | 2261 | UTR 3 | NP_899180.1 | |||
XM_011522719.2 | 2261 | UTR 3 | XP_011521021.1 | |||
XM_011522720.2 | 2261 | UTR 3 | XP_011521022.1 | |||
XM_011522721.1 | 2261 | Intron | XP_011521023.1 | |||
XM_011522722.1 | 2261 | Intron | XP_011521024.1 | |||
XM_011522723.1 | 2261 | Intron | XP_011521025.1 |