Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTGATAAAATCAAAGAATAGGTA[A/G]TGAAACAATCATCCAGTTAACAATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614901 MIM: 609912 MIM: 600823 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCKDK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BCKDK - branched chain ketoacid dehydrogenase kinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KAT8 - lysine acetyltransferase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032188.2 | Intron | NP_115564.2 | ||||
NM_182958.2 | Intron | NP_892003.2 | ||||
XM_011545970.1 | Intron | XP_011544272.1 | ||||
XM_011545971.1 | Intron | XP_011544273.1 |
PRSS8 - protease, serine 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |