Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACTGACCTTCAATGATGCCCCAC[C/T]TGGTCTTCCGGCCCAGGACACACCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611562 | ||||||||||||||||||||
Literature Links: |
SEPT12 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SEPT12 - septin 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001154458.2 | 1070 | Missense Mutation | NP_001147930.1 | |||
NM_144605.4 | 1070 | Missense Mutation | NP_653206.2 | |||
XM_006720846.2 | 1070 | Missense Mutation | XP_006720909.1 | |||
XM_011522379.2 | 1070 | Missense Mutation | XP_011520681.1 | |||
XM_017022938.1 | 1070 | Missense Mutation | XP_016878427.1 |
SMIM22 - small integral membrane protein 22 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |