Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTTTTAAATTTTTTTTCTTTTCC[C/T]GTGAGTTGGGAGTATTATGATGAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602698 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NFATC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NFATC3 - nuclear factor of activated T-cells 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004555.3 | Intron | NP_004546.1 | ||||
NM_173163.2 | Intron | NP_775186.1 | ||||
NM_173165.2 | Intron | NP_775188.1 |