Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCGTGGGGGTGGCCGACAGAGCA[A/G]TGGGGAAGGGGAACTGGCCCTCACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612081 MIM: 616951 | ||||||||||||||||||||
Literature Links: |
IL34 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IL34 - interleukin 34 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MTSS1L - metastasis suppressor 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138383.2 | 2563 | Missense Mutation | ACT,ATT | T,I 697 | NP_612392.1 | |
XM_005256236.4 | 2563 | Missense Mutation | ACT,ATT | T,I 795 | XP_005256293.2 | |
XM_011523450.2 | 2563 | Missense Mutation | ACT,ATT | T,I 741 | XP_011521752.1 | |
XM_011523453.2 | 2563 | Missense Mutation | ACT,ATT | T,I 849 | XP_011521755.2 | |
XM_011523454.2 | 2563 | Missense Mutation | ACT,ATT | T,I 846 | XP_011521756.2 | |
XM_011523458.2 | 2563 | Missense Mutation | ACT,ATT | T,I 568 | XP_011521760.1 | |
XM_017023879.1 | 2563 | Missense Mutation | ACT,ATT | T,I 817 | XP_016879368.1 | |
XM_017023880.1 | 2563 | Missense Mutation | ACT,ATT | T,I 805 | XP_016879369.1 | |
XM_017023881.1 | 2563 | Missense Mutation | ACT,ATT | T,I 798 | XP_016879370.1 | |
XM_017023882.1 | 2563 | Intron | XP_016879371.1 | |||
XM_017023883.1 | 2563 | Intron | XP_016879372.1 | |||
XM_017023884.1 | 2563 | Intron | XP_016879373.1 |