Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTGAATAACATGGCCCCTGCAAA[A/G]TAGTCTACTGAGTTCCTTAATCAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612147 MIM: 607213 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYLK3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MYLK3 - myosin light chain kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308301.1 | 2103 | UTR 3 | NP_001295230.1 | |||
NM_182493.2 | 2103 | UTR 3 | NP_872299.2 | |||
XM_006721334.3 | 2103 | UTR 3 | XP_006721397.1 |
ORC6 - origin recognition complex subunit 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |