Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTAAAAGCCTGTCATAAAAAAAAT[G/T]TAACAACTTTCTGCTTTAGTTTCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609604 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP1LC3B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAP1LC3B - microtubule associated protein 1 light chain 3 beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZCCHC14 - zinc finger CCHC-type containing 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015144.2 | 4468 | UTR 3 | NP_055959.1 | |||
XM_005255858.3 | 4468 | Intron | XP_005255915.2 | |||
XM_011522964.2 | 4468 | Intron | XP_011521266.1 | |||
XM_017023082.1 | 4468 | Intron | XP_016878571.1 |