Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGAATGTCACAAGCCCATGTGTCT[G/T]GGCAGCTGTCCGGCTTCAGTGCAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600182 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR6775 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR6775 - microRNA 6775 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC7A5 - solute carrier family 7 member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_006721286.3 | 1708 | UTR 3 | XP_006721349.2 | |||
XM_017023735.1 | 1708 | Intron | XP_016879224.1 | |||
XM_017023736.1 | 1708 | Intron | XP_016879225.1 | |||
XM_017023737.1 | 1708 | Intron | XP_016879226.1 |