Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCCCAGCTGGTGGTGTAGGGCGC[A/G]GAGGCTCTCCTGGTCCACAGTGCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611184 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTU2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CTU2 - cytosolic thiouridylase subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012759.2 | Intron | NP_001012777.1 | ||||
NM_001012762.2 | Intron | NP_001012780.1 | ||||
NM_001318507.1 | Intron | NP_001305436.1 | ||||
NM_001318513.1 | Intron | NP_001305442.1 | ||||
XM_017023210.1 | Intron | XP_016878699.1 |
MIR4722 - microRNA 4722 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PIEZO1 - piezo type mechanosensitive ion channel component 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF166 - ring finger protein 166 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |