Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATCTGGTCCAATCAAGTACATTA[C/T]TATTGTGTGATCCACCTGCAGAGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 160720 MIM: 603644 | ||||||||||||||||||||
Literature Links: |
MYH3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MYH3 - myosin, heavy chain 3, skeletal muscle, embryonic | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCO1 - SCO1 cytochrome c oxidase assembly protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004589.3 | 848 | Missense Mutation | ATA,GTA | I,V 263 | NP_004580.1 | |
XM_005256751.3 | 848 | Missense Mutation | ATA,GTA | I,V 152 | XP_005256808.1 |