Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGTGACAGGTCCATTGTCACGA[C/T]GTGTCCTGGTCCCCCTGGTTTTCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612077 MIM: 614218 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR22 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR22 - microRNA 22 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR22HG - MIR22 host gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TLCD2 - TLC domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164407.1 | 866 | Missense Mutation | CAT,CGT | H,R 247 | NP_001157879.1 |
WDR81 - WD repeat domain 81 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |