Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTCCTGAACTTCCTGGGAAGTCA[A/G]CCTTCTTCGGCACCTCAGATGAGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614906 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SNX11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SNX11 - sorting nexin 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013323.2 | 526 | Missense Mutation | ACC,GCC | T,A 87 | NP_037455.2 | |
NM_152244.1 | 526 | Missense Mutation | ACC,GCC | T,A 87 | NP_689450.1 | |
XM_005257260.3 | 526 | Missense Mutation | ACC,GCC | T,A 87 | XP_005257317.1 | |
XM_005257261.3 | 526 | Missense Mutation | ACC,GCC | T,A 79 | XP_005257318.1 | |
XM_005257262.3 | 526 | Missense Mutation | ACC,GCC | T,A 79 | XP_005257319.1 | |
XM_011524697.2 | 526 | Missense Mutation | ACC,GCC | T,A 87 | XP_011522999.1 |