Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATTGACAATAAGAGAAAACTGCT[C/T]TGTGTGCAGAGTATTTTCTCCTGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607833 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FLJ45513 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FLJ45513 - uncharacterized LOC729220 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TAC4 - tachykinin 4 (hemokinin) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077503.1 | Intron | NP_001070971.1 | ||||
NM_001077504.1 | Intron | NP_001070972.1 | ||||
NM_001077505.1 | Intron | NP_001070973.1 | ||||
NM_001077506.1 | Intron | NP_001070974.1 | ||||
NM_170685.2 | Intron | NP_733786.2 |