Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCACCGCCAAGGTCTGCCTCCCCA[A/G]CAAGACTGCCACCTGCTGGTCCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 106180 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACE PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACE - angiotensin I converting enzyme | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000789.3 | 513 | Missense Mutation | AAC,AGC | N,S 160 | NP_000780.1 | |
NM_001178057.1 | 513 | Intron | NP_001171528.1 | |||
NM_152830.2 | 513 | Intron | NP_690043.1 | |||
XM_006721737.3 | 513 | Intron | XP_006721800.2 |