Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGGCACCACAAAACTAGAAAGCA[A/G]TATCAGAGGTTCCAGAGCATCTGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612507 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCA9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABCA9 - ATP binding cassette subfamily A member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080283.3 | Intron | NP_525022.2 | ||||
XM_017024011.1 | Intron | XP_016879500.1 | ||||
XM_017024012.1 | Intron | XP_016879501.1 | ||||
XM_017024013.1 | Intron | XP_016879502.1 | ||||
XM_017024014.1 | Intron | XP_016879503.1 | ||||
XM_017024015.1 | Intron | XP_016879504.1 |