Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCCGCTCTGTCCCCAGCCCTGCA[A/G]CTGCTGCTGTGGCACAGTGCACTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138970 MIM: 607000 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSF3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CSF3 - colony stimulating factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000759.3 | 162 | Silent Mutation | CAA,CAG | Q,Q 16 | NP_000750.1 | |
NM_001178147.1 | 162 | Silent Mutation | CAA,CAG | Q,Q 16 | NP_001171618.1 | |
NM_172219.2 | 162 | Silent Mutation | CAA,CAG | Q,Q 16 | NP_757373.1 | |
NM_172220.2 | 162 | Silent Mutation | CAA,CAG | Q,Q 16 | NP_757374.2 |
MED24 - mediator complex subunit 24 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |