Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGTAAGTGAAATGATTGATGTAC[A/G]TTTTGTGGATATTTCATTTTCTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 158381 MIM: 613113 MIM: 164345 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EVI2B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EVI2B - ecotropic viral integration site 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006495.3 | 1061 | Missense Mutation | CGT,TGT | R,C 266 | NP_006486.3 | |
XM_005257946.3 | 1061 | Missense Mutation | CGT,TGT | R,C 281 | XP_005258003.1 |
NF1 - neurofibromin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000267.3 | 1061 | Intron | NP_000258.1 | |||
NM_001042492.2 | 1061 | Intron | NP_001035957.1 | |||
NM_001128147.2 | 1061 | Intron | NP_001121619.1 | |||
XM_005257983.2 | 1061 | Intron | XP_005258040.1 | |||
XM_005257984.2 | 1061 | Intron | XP_005258041.1 | |||
XM_006721922.2 | 1061 | Intron | XP_006721985.1 | |||
XM_006721924.2 | 1061 | Intron | XP_006721987.1 | |||
XM_006721925.2 | 1061 | Intron | XP_006721988.1 | |||
XM_006721926.3 | 1061 | Intron | XP_006721989.1 | |||
XM_011524852.2 | 1061 | Intron | XP_011523154.1 | |||
XM_011524857.2 | 1061 | Intron | XP_011523159.1 | |||
XM_017024689.1 | 1061 | Intron | XP_016880178.1 | |||
XM_017024690.1 | 1061 | Intron | XP_016880179.1 |
OMG - oligodendrocyte myelin glycoprotein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |