Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCTCTTATGGACAGAACTACAGC[A/G]GTTACTCCAGTTATGGACAAAGTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601574 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TAF15 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TAF15 - TATA-box binding protein associated factor 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003487.3 | 147 | Missense Mutation | AGT,GGT | S,G 52 | NP_003478.1 | |
NM_139215.2 | 147 | Missense Mutation | AGT,GGT | S,G 52 | NP_631961.1 | |
XM_011525315.1 | 147 | UTR 5 | XP_011523617.1 | |||
XM_017025181.1 | 147 | UTR 5 | XP_016880670.1 |