Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAAGCAGCTGTTGACTCACCTGAG[C/T]CCTTGTGGCCTTGCAGAATCAACGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614982 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CBX3P2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CBX3P2 - chromobox 3 pseudogene 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SMCHD1 - structural maintenance of chromosomes flexible hinge domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015295.2 | Intron | NP_056110.2 | ||||
XM_011525642.1 | Intron | XP_011523944.1 | ||||
XM_011525643.2 | Intron | XP_011523945.1 | ||||
XM_011525644.2 | Intron | XP_011523946.1 | ||||
XM_017025684.1 | Intron | XP_016881173.1 |