Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCAGGAGCCGGCTTTCTGTGTCAG[A/G]AGAGGCCACGCCAGCACTTTCTAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616131 MIM: 615766 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GACAT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GACAT2 - gastric cancer associated transcript 2 (non-protein coding) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MTCL1 - microtubule crosslinking factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015210.3 | Intron | NP_056025.2 | ||||
XM_005258098.3 | Intron | XP_005258155.1 | ||||
XM_005258099.4 | Intron | XP_005258156.1 | ||||
XM_011525640.2 | Intron | XP_011523942.1 | ||||
XM_011525641.2 | Intron | XP_011523943.1 | ||||
XM_017025671.1 | Intron | XP_016881160.1 | ||||
XM_017025672.1 | Intron | XP_016881161.1 | ||||
XM_017025673.1 | Intron | XP_016881162.1 | ||||
XM_017025674.1 | Intron | XP_016881163.1 | ||||
XM_017025675.1 | Intron | XP_016881164.1 |