Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCGCCCTAAGGGGTTTTCTTCTCC[C/G]AGGGAACCAGCGGGGAAACTGAGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603675 MIM: 602102 | ||||||||||||||||||||
Literature Links: |
LOC723805 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC723805 - interleukin-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPS16 - ribosomal protein S16 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SUPT5H - SPT5 homolog, DSIF elongation factor subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001111020.2 | 154 | Intron | NP_001104490.1 | |||
NM_001130824.1 | 154 | Intron | NP_001124296.1 | |||
NM_001130825.1 | 154 | Intron | NP_001124297.1 | |||
NM_001319990.1 | 154 | Intron | NP_001306919.1 | |||
NM_001319991.1 | 154 | Intron | NP_001306920.1 | |||
NM_003169.3 | 154 | UTR 5 | NP_003160.2 | |||
XM_017027174.1 | 154 | Intron | XP_016882663.1 |