Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTGTGGCAGCCATTAATGAGACT[C/T]TGTCTGGTTTCCATGGCAGGGACCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176395 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PSG6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PSG6 - pregnancy specific beta-1-glycoprotein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031850.3 | 1396 | Missense Mutation | AAG,GAG | K,E 422 | NP_001027020.1 | |
NM_002782.4 | 1396 | Intron | NP_002773.1 |