Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGATTTGGATGATGAAGACAGCA[A/G]TCCAAAGGGAAGGGAATTCAGGTAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 194555 | ||||||||||||||||||||
Literature Links: |
LOC100379224 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC100379224 - uncharacterized LOC100379224 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF224 - zinc finger protein 224 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321645.1 | 573 | Missense Mutation | ATC,GTC | I,V 72 | NP_001308574.1 | |
NM_013398.3 | 573 | Missense Mutation | ATC,GTC | I,V 72 | NP_037530.2 | |
XM_017027261.1 | 573 | Missense Mutation | ATC,GTC | I,V 72 | XP_016882750.1 |