Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCATCTCCATGATCTCTGGGCCAC[C/T]GGGGTACTCTTGTTGAATGGGGGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609857 MIM: 607548 | ||||||||||||||||||||
Literature Links: |
DMWD PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DMWD - dystrophia myotonica, WD repeat containing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RSPH6A - radial spoke head 6 homolog A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030785.3 | 2182 | Missense Mutation | AGT,GGT | S,G 671 | NP_110412.1 | |
XM_011527351.1 | 2182 | Intron | XP_011525653.1 |