Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGCCACTCACGGAAGTACGACTT[C/G]GACCAGCGTGTCTGCTGGCTCAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601463 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HAS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HAS1 - hyaluronan synthase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297436.1 | 1050 | Silent Mutation | TCC,TCG | S,S 379 | NP_001284365.1 | |
NM_001523.3 | 1050 | Silent Mutation | TCC,TCG | S,S 380 | NP_001514.2 | |
XM_011526884.2 | 1050 | UTR 3 | XP_011525186.1 |
SPACA6 - sperm acrosome associated 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316972.1 | 1050 | Intron | NP_001303901.1 | |||
NM_001316994.1 | 1050 | Intron | NP_001303923.1 |