Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAAATAAAGGGACAGTTAGAGATG[A/G]CCTTGCATTAAGGTGAGAACACAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF613 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF613 - zinc finger protein 613 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031721.3 | Intron | NP_001026891.2 | ||||
NM_024840.3 | Intron | NP_079116.2 | ||||
XM_005259269.3 | Intron | XP_005259326.1 | ||||
XM_011527333.2 | Intron | XP_011525635.1 | ||||
XM_017027315.1 | Intron | XP_016882804.1 |