Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGTGGATCGCATGATGATTAGTG[A/C]GGCTTGAACGCATACTAAAAGCTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 194558 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC107983997 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC107983997 - zinc finger protein 816-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF701 - zinc finger protein 701 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172655.1 | 1996 | Intron | NP_001166126.1 | |||
NM_018260.2 | 1996 | Intron | NP_060730.2 | |||
XM_011527092.1 | 1996 | Intron | XP_011525394.1 |
ZNF83 - zinc finger protein 83 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105549.1 | 1996 | Intron | NP_001099019.1 | |||
NM_001105550.1 | 1996 | Intron | NP_001099020.1 | |||
NM_001105551.1 | 1996 | Intron | NP_001099021.1 | |||
NM_001105552.1 | 1996 | Intron | NP_001099022.1 | |||
NM_001277945.1 | 1996 | Missense Mutation | CGC,CTC | R,L 472 | NP_001264874.1 | |
NM_001277946.1 | 1996 | Missense Mutation | CGC,CTC | R,L 472 | NP_001264875.1 | |
NM_001277947.1 | 1996 | Missense Mutation | CGC,CTC | R,L 472 | NP_001264876.1 | |
NM_001277948.1 | 1996 | Missense Mutation | CGC,CTC | R,L 472 | NP_001264877.1 | |
NM_001277949.1 | 1996 | Missense Mutation | CGC,CTC | R,L 472 | NP_001264878.1 | |
NM_001277951.1 | 1996 | Intron | NP_001264880.1 | |||
NM_001277952.1 | 1996 | Intron | NP_001264881.1 | |||
NM_018300.3 | 1996 | Intron | NP_060770.3 | |||
XM_017026951.1 | 1996 | Missense Mutation | CGC,CTC | R,L 472 | XP_016882440.1 |