Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTTGTACATTTCCTGGCCCAGGA[C/G]AGTCTCCCAGCACTGAAGAAGGAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604910 | ||||||||||||||||||||
Literature Links: |
CNOT3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CNOT3 - CCR4-NOT transcription complex subunit 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LENG1 - leukocyte receptor cluster member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMC4 - transmembrane channel like 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145303.2 | 1053 | Missense Mutation | CTC,GTC | L,V 479 | NP_001138775.2 | |
NM_144686.3 | 1053 | Missense Mutation | CTC,GTC | L,V 473 | NP_653287.2 | |
XM_011526486.2 | 1053 | Missense Mutation | CTC,GTC | L,V 319 | XP_011524788.1 |