Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGCCCGGAGAGCAGATGGGAGGT[A/G]CGGCGACAGTGTTTGACGAGAGCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613198 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COX6B2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COX6B2 - cytochrome c oxidase subunit 6B2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KMT5C - lysine methyltransferase 5C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032701.3 | 16 | UTR 5 | NP_116090.2 | |||
XM_005259338.3 | 16 | UTR 5 | XP_005259395.1 | |||
XM_006723442.3 | 16 | Intron | XP_006723505.1 | |||
XM_011527415.2 | 16 | Intron | XP_011525717.1 | |||
XM_017027388.1 | 16 | Intron | XP_016882877.1 |
TMEM150B - transmembrane protein 150B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |