Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGGATCTTCTGAGTTTCTTCATC[A/G]AAGCCAGATTTGTGCAGCCTGGGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609663 | ||||||||||||||||||||
Literature Links: |
NLRP9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NLRP9 - NLR family pyrin domain containing 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_176820.3 | 922 | Silent Mutation | TTC,TTT | F,F 954 | NP_789790.2 | |
XM_011526894.2 | 922 | Silent Mutation | TTC,TTT | F,F 242 | XP_011525196.1 | |
XM_011526895.1 | 922 | Silent Mutation | TTC,TTT | F,F 242 | XP_011525197.1 |