Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGAAAAAGCTGAAAGTATGATGT[A/G]TAGAATAGGTGGAACCCATTAGCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF583 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF583 - zinc finger protein 583 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159860.1 | Intron | NP_001153332.1 | ||||
NM_001159861.1 | Intron | NP_001153333.1 | ||||
NM_152478.2 | Intron | NP_689691.2 | ||||
XM_005258558.1 | Intron | XP_005258615.1 | ||||
XM_011526517.2 | Intron | XP_011524819.1 | ||||
XM_017026346.1 | Intron | XP_016881835.1 | ||||
XM_017026347.1 | Intron | XP_016881836.1 | ||||
XM_017026348.1 | Intron | XP_016881837.1 | ||||
XM_017026349.1 | Intron | XP_016881838.1 | ||||
XM_017026350.1 | Intron | XP_016881839.1 | ||||
XM_017026351.1 | Intron | XP_016881840.1 | ||||
XM_017026352.1 | Intron | XP_016881841.1 | ||||
XM_017026353.1 | Intron | XP_016881842.1 |