Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCTCACTCAGGACAGAGGGCTCT[A/G]ATCTAGGACCAGCAACTCACACCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606834 MIM: 605859 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KMT2B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KMT2B - lysine methyltransferase 2B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZBTB32 - zinc finger and BTB domain containing 32 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316902.1 | 582 | Intron | NP_001303831.1 | |||
NM_001316903.1 | 582 | Intron | NP_001303832.1 | |||
NM_014383.2 | 582 | Intron | NP_055198.1 | |||
XM_011526718.2 | 582 | Intron | XP_011525020.1 | |||
XM_017026588.1 | 582 | Intron | XP_016882077.1 | |||
XM_017026589.1 | 582 | UTR 5 | XP_016882078.1 | |||
XM_017026590.1 | 582 | Intron | XP_016882079.1 | |||
XM_017026591.1 | 582 | Intron | XP_016882080.1 | |||
XM_017026592.1 | 582 | Intron | XP_016882081.1 |