Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACCTCACTGGGCGAGGGCCAGCT[A/G]CGCGGCCTGGTCAACTTGCGCCACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612809 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105372383 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105372383 - uncharacterized LOC105372383 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRFN3 - leucine rich repeat and fibronectin type III domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024509.1 | 833 | Silent Mutation | CTA,CTG | L,L 127 | NP_078785.1 | |
XM_017027302.1 | 833 | Silent Mutation | CTA,CTG | L,L 127 | XP_016882791.1 |