Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCCATGGGCTGGCCTGCATAGAC[A/G]GGTCTGAGGGGCATGGGCCTTTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603619 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FZR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FZR1 - fizzy/cell division cycle 20 related 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136197.1 | Intron | NP_001129669.1 | ||||
NM_001136198.1 | Intron | NP_001129670.1 | ||||
NM_016263.3 | Intron | NP_057347.2 | ||||
XM_005259573.4 | Intron | XP_005259630.1 | ||||
XM_017026863.1 | Intron | XP_016882352.1 |