Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCAGTTGTAGGAGACTTTCTCCT[C/T]AGCCTCCACGAACAAGGGGTTCTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARRDC5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARRDC5 - arrestin domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080523.1 | 257 | Missense Mutation | AAG,GAG | K,E 175 | NP_001073992.1 | |
XM_005259616.2 | 257 | Missense Mutation | AAG,GAG | K,E 37 | XP_005259673.1 | |
XM_011528181.2 | 257 | Missense Mutation | AAG,GAG | K,E 37 | XP_011526483.1 | |
XM_017027144.1 | 257 | Intron | XP_016882633.1 | |||
XM_017027145.1 | 257 | Intron | XP_016882634.1 |