Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTCAGATGTTAAGACTGTGTTCA[C/T]CCAGCCCGACTTTCCTTTCTTTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC00664 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LINC00664 - long intergenic non-protein coding RNA 664 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF429 - zinc finger protein 429 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001415.2 | Intron | NP_001001415.2 | ||||
XM_017026748.1 | Intron | XP_016882237.1 | ||||
XM_017026749.1 | Intron | XP_016882238.1 | ||||
XM_017026750.1 | Intron | XP_016882239.1 | ||||
XM_017026751.1 | Intron | XP_016882240.1 | ||||
XM_017026752.1 | Intron | XP_016882241.1 | ||||
XM_017026753.1 | Intron | XP_016882242.1 | ||||
XM_017026754.1 | Intron | XP_016882243.1 | ||||
XM_017026755.1 | Intron | XP_016882244.1 | ||||
XM_017026756.1 | Intron | XP_016882245.1 | ||||
XM_017026757.1 | Intron | XP_016882246.1 |