Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTGGTCCATAGTTCCAGAATTCA[C/T]TTCAGGATCAGAGATGTGGTGAACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604350 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAB3D PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RAB3D - RAB3D, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004283.3 | 1669 | UTR 3 | NP_004274.1 |
TSPAN16 - tetraspanin 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282509.1 | 1669 | Intron | NP_001269438.1 | |||
NM_001282510.1 | 1669 | Intron | NP_001269439.1 | |||
NM_012466.3 | 1669 | Intron | NP_036598.1 | |||
XM_011527901.1 | 1669 | Intron | XP_011526203.1 | |||
XM_011527902.1 | 1669 | Intron | XP_011526204.1 | |||
XM_011527903.2 | 1669 | Intron | XP_011526205.1 |