Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGCTCGGCCACCAACCTGAGCCG[C/T]GTGGCGGGCCTGGAGAAGCAGTTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601032 | ||||||||||||||||||||
Literature Links: |
PKN1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PKN1 - protein kinase N1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002741.3 | 416 | Silent Mutation | CGC,CGT | R,R 127 | NP_002732.3 | |
NM_213560.1 | 416 | Silent Mutation | CGC,CGT | R,R 133 | NP_998725.1 | |
XM_011528128.1 | 416 | Silent Mutation | CGC,CGT | R,R 139 | XP_011526430.1 |