Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCCATGTAGGCCTCCTTCTCACC[A/G]AACCTGGATCAGGGCCAGGCTGGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616561 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR1470 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR1470 - microRNA 1470 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RASAL3 - RAS protein activator like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022904.1 | 2039 | Silent Mutation | TTC,TTT | F,F 654 | NP_075055.1 | |
XM_011528185.1 | 2039 | Silent Mutation | TTC,TTT | F,F 657 | XP_011526487.1 | |
XM_011528186.1 | 2039 | Silent Mutation | TTC,TTT | F,F 651 | XP_011526488.1 | |
XM_011528187.1 | 2039 | Silent Mutation | TTC,TTT | F,F 657 | XP_011526489.1 |
WIZ - widely interspaced zinc finger motifs | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |