Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATCTGCCGGAACTACCGTGGCGA[C/T]GTGGACATGTCAGAGGTGGAGCACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603535 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AP1M1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AP1M1 - adaptor related protein complex 1 mu 1 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130524.1 | 245 | Silent Mutation | GAC,GAT | D,D 24 | NP_001123996.1 | |
NM_032493.3 | 245 | Silent Mutation | GAC,GAT | D,D 24 | NP_115882.1 | |
XM_017027420.1 | 245 | UTR 5 | XP_016882909.1 | |||
XM_017027421.1 | 245 | UTR 5 | XP_016882910.1 |