Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACACACGGGGATACAGGCTGGCA[C/G]CCCAGTTCCCACAGTCTGGGCTGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612436 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLCO4A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLCO4A1 - solute carrier organic anion transporter family member 4A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016354.3 | Intron | NP_057438.3 | ||||
XM_005260203.2 | Intron | XP_005260260.1 | ||||
XM_011528792.2 | Intron | XP_011527094.1 | ||||
XM_017027826.1 | Intron | XP_016883315.1 | ||||
XM_017027827.1 | Intron | XP_016883316.1 |