Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCTCTGGCCTCCTGATCGAGAGA[C/T]ACCAAGGAAGAAAAAAGCTGAATCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR3617 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR3617 - microRNA 3617 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WFDC10B - WAP four-disulfide core domain 10B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_172006.2 | Intron | NP_742003.1 | ||||
NM_172131.2 | Intron | NP_742143.1 | ||||
XM_017027824.1 | Intron | XP_016883313.1 | ||||
XM_017027825.1 | Intron | XP_016883314.1 |
WFDC13 - WAP four-disulfide core domain 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_172005.1 | Intron | NP_742002.1 |
WFDC3 - WAP four-disulfide core domain 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |