Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACTGGCAGGGACTCAGCTGAAGT[C/T]GCTGCTCTCCCTCACCCCTGCTAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616825 MIM: 606726 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NCOA5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NCOA5 - nuclear receptor coactivator 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020967.2 | 3001 | UTR 3 | NP_066018.1 | |||
XM_005260474.3 | 3001 | UTR 3 | XP_005260531.1 | |||
XM_011528951.2 | 3001 | UTR 3 | XP_011527253.1 | |||
XM_017027988.1 | 3001 | UTR 3 | XP_016883477.1 |
SLC12A5 - solute carrier family 12 member 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |