Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTTGCCTTTGGTGGTAAAAGCTC[G/A]CCCACAAATGTTGCACACAAAAGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607343 | ||||||||||||||||||||
Literature Links: |
SALL4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SALL4 - spalt like transcription factor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318031.1 | 3004 | Nonsense Mutation | CGA,TGA | R,* 468 | NP_001304960.1 | |
NM_020436.4 | 3004 | Nonsense Mutation | CGA,TGA | R,* 905 | NP_065169.1 | |
XM_005260467.3 | 3004 | Intron | XP_005260524.1 | |||
XM_011528921.2 | 3004 | Nonsense Mutation | CGA,TGA | R,* 803 | XP_011527223.1 | |
XM_011528922.2 | 3004 | Nonsense Mutation | CGA,TGA | R,* 803 | XP_011527224.1 |