Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTTTGCTTTGCTTTCGTCACTGT[C/T]GCCCAGCCATCGCCTCTGAAGGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZFP64 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZFP64 - ZFP64 zinc finger protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319146.1 | 2081 | UTR 3 | NP_001306075.1 | |||
NM_018197.2 | 2081 | Intron | NP_060667.2 | |||
NM_022088.4 | 2081 | Intron | NP_071371.3 | |||
NM_199426.1 | 2081 | Intron | NP_955458.1 | |||
NM_199427.2 | 2081 | UTR 3 | NP_955459.2 | |||
XM_005260449.1 | 2081 | Intron | XP_005260506.1 | |||
XM_006723822.1 | 2081 | Intron | XP_006723885.1 | |||
XM_011528900.1 | 2081 | Intron | XP_011527202.1 | |||
XM_017027945.1 | 2081 | UTR 3 | XP_016883434.1 | |||
XM_017027946.1 | 2081 | Intron | XP_016883435.1 | |||
XM_017027947.1 | 2081 | UTR 3 | XP_016883436.1 | |||
XM_017027948.1 | 2081 | Intron | XP_016883437.1 | |||
XM_017027949.1 | 2081 | Intron | XP_016883438.1 |