Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTAAAGAAAAAAAGAAATGCTTCC[C/T]CCAACAAAGACTGTGCATTCAATAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM209A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM209A - family with sequence similarity 209 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012971.3 | 399 | Missense Mutation | CCC,TCC | P,S 107 | NP_001012989.2 |
FAM209B - family with sequence similarity 209 member B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GCNT7 - glucosaminyl (N-acetyl) transferase family member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080615.1 | 399 | UTR 5 | NP_542182.1 |
RTFDC1 - replication termination factor 2 domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |