Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCTGCCCGCTGGCCCCGTGCAGC[A/T]GCACAGGGTGGAGGCTGGGGTGGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614425 | ||||||||||||||||||||
Literature Links: |
TTI1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TTI1 - TELO2 interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303457.1 | 3303 | Missense Mutation | CAG,CTG | Q,L 1066 | NP_001290386.1 | |
NM_014657.2 | 3303 | Missense Mutation | CAG,CTG | Q,L 1066 | NP_055472.1 | |
XM_011529114.1 | 3303 | Intron | XP_011527416.1 | |||
XM_017028148.1 | 3303 | Intron | XP_016883637.1 |