Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAAGTGCTATTTGTAACTTAGAT[A/T]GAATGTTGAAACTTGTGTTTGTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602908 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
POLR3F PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
POLR3F - RNA polymerase III subunit F | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RBBP9 - RB binding protein 9, serine hydrolase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006606.2 | 772 | UTR 3 | NP_006597.2 | |||
XM_005260652.2 | 772 | UTR 3 | XP_005260709.1 |