Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCAGTAAATGCGACGATTAGAAGC[C/T]TCTGCTTCCGTTTTCCATCTGGACG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600687 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TIAM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TIAM1 - T-cell lymphoma invasion and metastasis 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003253.2 | 7028 | UTR 3 | NP_003244.2 | |||
XM_005261037.1 | 7028 | UTR 3 | XP_005261094.1 | |||
XM_005261038.2 | 7028 | UTR 3 | XP_005261095.1 | |||
XM_005261040.2 | 7028 | UTR 3 | XP_005261097.1 | |||
XM_011529711.1 | 7028 | UTR 3 | XP_011528013.1 | |||
XM_011529712.1 | 7028 | UTR 3 | XP_011528014.1 | |||
XM_011529713.1 | 7028 | UTR 3 | XP_011528015.1 | |||
XM_017028447.1 | 7028 | UTR 3 | XP_016883936.1 | |||
XM_017028448.1 | 7028 | UTR 3 | XP_016883937.1 | |||
XM_017028449.1 | 7028 | UTR 3 | XP_016883938.1 | |||
XM_017028450.1 | 7028 | UTR 3 | XP_016883939.1 | |||
XM_017028451.1 | 7028 | UTR 3 | XP_016883940.1 | |||
XM_017028452.1 | 7028 | UTR 3 | XP_016883941.1 | |||
XM_017028453.1 | 7028 | UTR 3 | XP_016883942.1 |